Learn More
HCL Labels 32 oz Pre-labeled GHS Spray Bottle Zep All-Purpose Cleaner, Bottle Supplied Empty, 1 Ea
SDP
Catalog No. 501633409
Encompass_Preferred
Bottles, HCL Labels, Inc., 32oz, GHS Zep All-purpose Cleaner Spray Bottle, These bottles are pre-labeled with a GHS hazard communication chemical label. They are fully compliant and include the necessary pictograms and signal words. Bottles are chemical resistant and empty.
Catalog No. 50-163-3409 Supplier HCL Labels Supplier No. GHSBOT0054
Bottles, HCL Labels, Inc., 32oz, GHS Zep All-purpose Cleaner Spray Bottle, These bottles are pre-labeled with a GHS hazard communication chemical label. They are fully compliant and include the necessary pictograms and signal words. Bottles are chemical resistant and empty.