Learn More
Thermco Products Inc Digital Refrigerator Thermometer Thermco Fahrenheit / Celsius -58° to +158°F (-50° to +70°C) Glycol Bottle Probe Door / Wall Mount Battery Operated
Catalog No. NC1023610
Encompass_Preferred
Lollipop digital bottle thermometer. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend. NIST Traceable. Certificate provided with critical temperature certified. Current temperatures with minimum/maximum function. Records the high & low temperature over time period.Each thermometer individually serialized.
Catalog No. NC1023610 Supplier Thermco Products Inc Supplier No. ACC304REFL
Lollipop digital bottle thermometer. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend. NIST Traceable. Certificate provided with critical temperature certified. Current temperatures with minimum/maximum function. Records the high & low temperature over time period.Each thermometer individually serialized.