Learn More
COLE PARMER INSTRUMENT COMPANY BOTTLE THREAD ADAPTER 38-430 F
Catalog No. NC1604783
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
BOTTLE THREAD ADAPTER 38-430 female to GL45 male Polypropylene
Catalog No. NC1604783 Supplier COLE PARMER INSTRUMENT COMPANY Supplier No. 1202251
This item has a minimum qty of 2 per supplier requirements.
BOTTLE THREAD ADAPTER 38-430 female to GL45 male Polypropylene