Learn More
Electron Microscopy Sciences Bottle Top Unit 1000 ml Funnel Tubing, CA, 45 mm neck, 0.22 µm pores

Supplier: Electron Microscopy Sciences 6705254
Bottle-top unit for use with disposable sterile vacuum filter. Connector neck with unique polyethylene gasket ensures a positive seal between filtration unit and collecting bottle. Funnel has tubing adapter that fits most vacuum hoses. Units come individually packaged and are pre-sterilized by gamma irradiation, certified non-pyrogenic. Prefilters included.
Membranes are detergent-free, tissue culture compatible, and heat-sealed to the support grid to maximize flow rate, reduce foaming and protein denaturization. Ideal for filtration of tissue culture media, biological fluids, fixation buffers etc. (not for parenteral use).
Material: CA
Neck: 45 mm
Pore size: 0.22 µm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.