Learn More
Labsciences BOTTLE TOP DISPENSER 0.5-5 ML
Catalog No. NC0177790
Encompass_Preferred
Smooth plunger movement for accurate and precise dispensing. Fully autoclavable and compatible with all reagents except HF. Easy to disassemble for cleaning and servicing. Includes 5 adapters that fit most reagent bottles.
Catalog No. NC0177790 Supplier Labsciences Supplier No. 5000BTR
Smooth plunger movement for accurate and precise dispensing. Fully autoclavable and compatible with all reagents except HF. Easy to disassemble for cleaning and servicing. Includes 5 adapters that fit most reagent bottles.