Learn More
JDM LAB SOLUTIONS LLC ACCUTOP BOTTLE TOP DISPENSER - 5-60 ml,
Catalog No. NC3706395
Encompass
AccuTop Bottle Top Dispenser, Without Re-Circulation 5-60 ml, ergonomically designed bottle top dispenser that is ideal for quick and easy use.
Catalog No. NC3706395 Supplier JDM LAB SOLUTIONS LLC Supplier No. AD60
AccuTop Bottle Top Dispenser, Without Re-Circulation 5-60 ml, ergonomically designed bottle top dispenser that is ideal for quick and easy use.