Learn More
JDM LAB SOLUTIONS LLC ACCUTOP BOTTLE TOP DISPENSER -0.5-5 ml,
Catalog No. NC3706396
Encompass
AccuTop Bottle Top Dispenser, With Re-Circulation 0.5-5 ml, ergonomically designed bottle top dispenser that is ideal for quick and easy use.
Catalog No. NC3706396 Supplier JDM LAB SOLUTIONS LLC Supplier No. ADR05
AccuTop Bottle Top Dispenser, With Re-Circulation 0.5-5 ml, ergonomically designed bottle top dispenser that is ideal for quick and easy use.