Learn More
Electron Microscopy Sciences TorrLube Bottle Torrlube Oil 60 CM3
SDP
Catalog No. 5034119
Encompass_Preferred
TorrLube Oil is an extremely pure PFPE (Perfluoropolyalkether ) blend which is thoroughly distilled and then filtered through a submicron screen. TorrLube Oil has the ability to withstand thermal expansion and contraction as well as chemical and mechanical stress, while still maintaining consistent viscosity. These properties make TorrLube the ideal lubricant for bearings, automated slides, and screws that have a tendency to gall or cold weld while working under extreme conditions.
A set of two small bottles, each containing 60cc of TorrLube lubricant. Every bottle comes with a sealed cap. A luer lock cap and a #22 gauge blunt needle are provided for closed application. The bottle set is appropriate for larger manufacturing units and production fabs. Each twin pack of TorrLube 60cc bottles is sealed to prevent leakage and contamination.
Catalog No. 50-341-19 Supplier Electron Microscopy Sciences Supplier No. 6075201
TorrLube Oil is an extremely pure PFPE (Perfluoropolyalkether ) blend which is thoroughly distilled and then filtered through a submicron screen. TorrLube Oil has the ability to withstand thermal expansion and contraction as well as chemical and mechanical stress, while still maintaining consistent viscosity. These properties make TorrLube the ideal lubricant for bearings, automated slides, and screws that have a tendency to gall or cold weld while working under extreme conditions.
A set of two small bottles, each containing 60cc of TorrLube lubricant. Every bottle comes with a sealed cap. A luer lock cap and a #22 gauge blunt needle are provided for closed application. The bottle set is appropriate for larger manufacturing units and production fabs. Each twin pack of TorrLube 60cc bottles is sealed to prevent leakage and contamination.