Learn More
Hach Company Presterilized Sampling Bottle, Round, 50 per pack
Supplier:  Hach Company 2495050
Encompass
Polystyrene, disposable, transparent, non-fluorescent, break-resistant, presterilized bottles with caps. Bottles have 100 mL fill-to line; capacity of 175 mL. Bottles used with medium containing MUG or other fluorescing reagent. Work well with P-A Broth Ampules and PathoScreen™ Medium.
Catalog No. 50-199-6076