Learn More
Hach Company Bottle, Storage, Polyethylene, Round, Amber, 500 mL, 6/pk
Catalog No. 501995256 Shop All Hach Company Products
Encompass
High-density polyethylene bottle with leak-proof polypropylene cap. Ideal for storage of light sensitive materials, solid or liquid.
Catalog No. 50-199-5256 Supplier Hach Company Supplier No. 2085859
High-density polyethylene bottle with leak-proof polypropylene cap. Ideal for storage of light sensitive materials, solid or liquid.