Learn More
Hach Company Bottle, Storage, Polyethylene, Rectangular, 2000 mL, 4/pk
Catalog No. 501995270 Shop All Hach Company Products
Encompass
Heavy-walled, high-density polyethylene (HDPE) bottle features wide mouth for easy filling and leak-proof polypropylene cap. Rectangular shape for maximum space efficiency and less damage in shipping. Suitable for both aqueous solutions and solid samples. Amber color bottle decreases photo degradation by sunlight.
Catalog No. 50-199-5270 Supplier Hach Company Supplier No. 2086784
Heavy-walled, high-density polyethylene (HDPE) bottle features wide mouth for easy filling and leak-proof polypropylene cap. Rectangular shape for maximum space efficiency and less damage in shipping. Suitable for both aqueous solutions and solid samples. Amber color bottle decreases photo degradation by sunlight.