Learn More
Hach Company Bottle, Sampling, Polycarbonate, 500 mL, 4/pk
Catalog No. 501995808 Shop All Hach Company Products
Encompass
Transparent polycarbonate bottle with cap features wide mouth (3.8cm/1.48" diameter) opening for easy filling and square design for maximum space efficiency. Autoclavable with molded-in graduations (do not engage threads for autoclave cycle.)
Catalog No. 50-199-5808 Supplier Hach Company Supplier No. 2324539
Transparent polycarbonate bottle with cap features wide mouth (3.8cm/1.48" diameter) opening for easy filling and square design for maximum space efficiency. Autoclavable with molded-in graduations (do not engage threads for autoclave cycle.)