Learn More
Hach Company Bottle, Storage, Polyethylene, Square, 175 mL, 12/pk
Supplier:  Hach Company 2086874
Encompass
High-density polyethylene bottle with leak-proof polypropylene cap. Square design saves space, while wide-mouth allows for easy fill and removal of contents.
Catalog No. 50-199-5272