Learn More
Hach Company Bottle, Sampling, Low Density, Polyethylene, 1 L, 6/pk
Supplier:  Hach Company 2087083
Encompass
Low-density polyethylene (LDPE) bottles feature wide-mouth opening for easy filling and leak-proof polypropylene closure. Great choice for grab samples, shipping liquids, soil sampling, and storing dry chemicals.
Catalog No. 50-199-5281