Learn More
Hach Company Laboratory Bottles, Polypropylene, Autoclavable, Wide Mouth, 1-L, 6/pk
Catalog No. 501995103 Shop All Hach Company Products
Encompass
Polypropylene bottles suitable for sample collection to chemical storage. Matched polypropylene caps have a valve-seal design for a perfect, leakproof fit. A protruding ring under the cap holds shrink-wrapping securely in place. Autoclavable 121°C (Do not autoclave with caps in place). Available in Narrow Mouth or Wide Mouth.
Catalog No. 50-199-5103 Supplier Hach Company Supplier No. 1472463
Polypropylene bottles suitable for sample collection to chemical storage. Matched polypropylene caps have a valve-seal design for a perfect, leakproof fit. A protruding ring under the cap holds shrink-wrapping securely in place. Autoclavable 121°C (Do not autoclave with caps in place). Available in Narrow Mouth or Wide Mouth.