Learn More
Hach Company Laboratory Bottles, Polypropylene, Autoclavable, Wide Mouth, 500 mL, 12/pk
Catalog No. 501995102 Shop All Hach Company Products
Encompass
Polyepropylene bottles suitable for sample collection to chemical storage. Matched polypropylene caps have a valve-seal design for a perfect, leakproof fit. A protruding ring under the cap holds shrink-wrapping securely in place. Autoclavable 121°C (Do not autoclave with caps in place). Available in Narrow Mouth or Wide Mouth.
Catalog No. 50-199-5102 Supplier Hach Company Supplier No. 1472459
Polyepropylene bottles suitable for sample collection to chemical storage. Matched polypropylene caps have a valve-seal design for a perfect, leakproof fit. A protruding ring under the cap holds shrink-wrapping securely in place. Autoclavable 121°C (Do not autoclave with caps in place). Available in Narrow Mouth or Wide Mouth.