Learn More
Hach Company Bottle, BOD, 300 mL, 24/pk
Supplier:  Hach Company 62124
Encompass
The standard for BOD analysis. Borosilicate glass bottle with flared lip and ground-glass stopper to prevent air entrapment and for water seal. Frosted marking area. Stopper included (60 and 300 mL sizes). Serialized bottles have silk-screened, sequential numbers and include a bar code.
Catalog No. 50-199-4414