Learn More
Hach Company Bottle, Wash, Polyethylene, 1000 mL
Supplier:  Hach Company 62016
Encompass
Low-density polyethylene bottle with polypropylene screw closure features tubulation molded into bottle to eliminate any possibility of leakage. Entire contents can be dispensed without tipping or shaking.
Catalog No. 50-199-4409