Learn More
Hach Company Bottle, Wash, Autoclavable, 1000 mL, PK/4
Supplier:  Hach Company 2930202
Encompass
Polypropylene copolymer bottle with specially molded polypropylene screw closure for leak-proof service. Autoclavable. Before autoclaving, set cap on top of bottle without engaging threads.
Catalog No. 50-199-6837