Learn More
Hach Company Bottle, Wash, Polyethylene, Narrow Mouth, 250 mL, 12/pk
Catalog No. 501996835 Shop All Hach Company Products
Encompass
Polyethylene bottle with polypropylene closure and stem molded in one piece for leak-proof service. Tip can be cutback to increase flow. Available in Narrow or Wide Mouth.
Catalog No. 50-199-6835 Supplier Hach Company Supplier No. 2930102
Polyethylene bottle with polypropylene closure and stem molded in one piece for leak-proof service. Tip can be cutback to increase flow. Available in Narrow or Wide Mouth.