Learn More
Hach Company Bottle, Wash, Polyethylene, Narrow Mouth, 500 mL, 12/pk
Supplier:  Hach Company 2930103
Encompass
Polyethylene bottle with polypropylene closure and stem molded in one piece for leak-proof service. Tip can be cutback to increase flow. Available in Narrow or Wide Mouth.
Catalog No. 50-199-6836