Learn More
Hach Company Bottle-wash-PE-125 mL
Supplier:  Hach Company 62014
Encompass
Low-density polyethylene bottle with polypropylene screw closure features tubulation molded into bottle to eliminate any possibility of leakage Entire contents can be dispensed without tipping or shaking
Catalog No. 50-002-57107