Learn More
Hach Company Bottle-Wash-PE-750 mL
Supplier:  Hach Company 62075
Encompass
Low-density polyethylene bottle with polypropylene screw closure features tubulation molded into bottle to eliminate any possibility of leakage Entire contents can be dispensed without tipping or shaking
Catalog No. 50-002-57159