Learn More
Hach Company Bottle, Wash, Diamond Alert, 500 mL, 6/pk
Supplier:  Hach Company 2613049
Encompass
Wide-mouthed and color-coded wash bottles provide safe and positive identification of common laboratory solutions. Standard Diamond Alert symbols indicate type of hazard contents may represent. Polyethylene bottles with color-coded polypropylene screw caps and spray-tip assembly. Variety 6-pack includes one bottle each for water, acetone, methanol, isopropanol, toluene, and ethanol.
Catalog No. 50-199-6351