Learn More
Waters Corp Waters Waste Bottle Valve Assembly, 48VDC, MS Systems
Catalog No. 50466679 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement waste bottle valve assembly includes a 48V valve. It is designed for use with SQD2, Xevo TQD, and Xevo TQ-S micro systems.
  • Replacement source exhaust waste bottle with a 48V valve.
  • Designed for use with SQD2, Xevo TQD, and Xevo TQ-S micro systems.
  • Spare parts kit.
  • Valve type component.
  • For MS system type.
  • Replacement as required.
Catalog No. 50-466-679 Supplier Waters Corp Supplier No. 700005811
This replacement waste bottle valve assembly includes a 48V valve. It is designed for use with SQD2, Xevo TQD, and Xevo TQ-S micro systems.
  • Replacement source exhaust waste bottle with a 48V valve.
  • Designed for use with SQD2, Xevo TQD, and Xevo TQ-S micro systems.
  • Spare parts kit.
  • Valve type component.
  • For MS system type.
  • Replacement as required.