Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Graduated Wide Mouth LD Polyethylene Bottle 1000ml, 206 mm height, 55 mm I.D., 95 mm O.D.
SDP

Catalog No. 501928753
Encompass_Preferred

These Low-density polyethylene bottles are flexible and feature a wide-mouth opening for easy filling and emptying of liquids and powders. The deep threaded neck allows the screw cap to seal on tight, offering exceptional leak-resistance, especially during shipping. This bottle is approved for use with food products. Flexible bottle is made from low-density polyethylene and the cap is made from polypropylene. Graduated.
Capacity: 1000 ml
Height: 206 mm
I.D.: 55 mm
O.D.: 95 mm
Gradations: 100 ml

Catalog No. 50-192-8753 Supplier Electron Microscopy Sciences Supplier No. 6424210
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Low-density polyethylene bottles are flexible and feature a wide-mouth opening for easy filling and emptying of liquids and powders. The deep threaded neck allows the screw cap to seal on tight, offering exceptional leak-resistance, especially during shipping. This bottle is approved for use with food products. Flexible bottle is made from low-density polyethylene and the cap is made from polypropylene. Graduated.
Capacity: 1000 ml
Height: 206 mm
I.D.: 55 mm
O.D.: 95 mm
Gradations: 100 ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.