Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Graduated Wide Mouth LD Polyethylene Bottle 2000ml, 252 mm height, 55 mm I.D., 120 mm O.D.
SDP

Catalog No. 501928755
Encompass_Preferred

These Low-density polyethylene bottles are flexible and feature a wide-mouth opening for easy filling and emptying of liquids and powders. The deep threaded neck allows the screw cap to seal on tight, offering exceptional leak-resistance, especially during shipping. This bottle is approved for use with food products. Flexible bottle is made from low-density polyethylene and the cap is made from polypropylene. Graduated.
Capacity: 2000 ml
Height: 252 mm
I.D.: 55 mm
O.D.: 120 mm
Gradations: 100 ml

Catalog No. 50-192-8755 Supplier Electron Microscopy Sciences Supplier No. 6424212
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Low-density polyethylene bottles are flexible and feature a wide-mouth opening for easy filling and emptying of liquids and powders. The deep threaded neck allows the screw cap to seal on tight, offering exceptional leak-resistance, especially during shipping. This bottle is approved for use with food products. Flexible bottle is made from low-density polyethylene and the cap is made from polypropylene. Graduated.
Capacity: 2000 ml
Height: 252 mm
I.D.: 55 mm
O.D.: 120 mm
Gradations: 100 ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.