Learn More
Hach Company Bottle, Sampling, High Density, Polyethylene, 2 Liter
Supplier:  Hach Company 2087154
Encompass
High-density polyethylene bottle with wide-mouth (89 mm) opening. Excellent sampling bottle for grab samples, storing dry chemicals, and shipping liquids.
Catalog No. 50-199-5283