Learn More
Cole-Parmer Essentials Wide-Mouth Plastic Bottle, PP, 1000mL (32 oz); 24/CS
Catalog No. NC2066667 Shop All Cole-Parmer Products
Encompass_Preferred
Manufactured from USP Class VI virgin polypropylene (PP) resin, these wide mouth bottles offer excellent chemical resistance and clarity. Precision molded for consistent wall thickness and smooth interior and exterior sufaces. The wide mouth opening is perfect for sample collection and allows easy filling and emptying of solids, powders, and viscous liquids.
Catalog No. NC2066667 Supplier Cole-Parmer Supplier No. 0604551
Manufactured from USP Class VI virgin polypropylene (PP) resin, these wide mouth bottles offer excellent chemical resistance and clarity. Precision molded for consistent wall thickness and smooth interior and exterior sufaces. The wide mouth opening is perfect for sample collection and allows easy filling and emptying of solids, powders, and viscous liquids.