Learn More
Hach Company Bottle, Storage, Autoclavable, Wide Mouth, 125 mL pk/12
Supplier:  Hach Company 2939103
Encompass
Polypropylene bottles suitable for sample collection to chemical storage. Matched polypropylene caps have a valve-seal design for a perfect, leakproof fit. A protruding ring under the cap holds shrink-wrapping securely in place. Autoclavable 121°C (Do not autoclave with caps in place). Available in Narrow Mouth or Wide Mouth.
Catalog No. 50-199-6862