Learn More
Med Vet International Yorker Dropper Bottle, 15mL, 12/Box
Catalog No. 502522484
Encompass_Preferred
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene.
Catalog No. 50-252-2484 Supplier Med Vet International Supplier No. 975015
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene.