Learn More
Med Vet International Yorker Dropper Bottle, 30mL, 12/Box
Catalog No. 502522483
Encompass_Preferred
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene. 12 per box.
Catalog No. 50-252-2483 Supplier Med Vet International Supplier No. 975030
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene. 12 per box.