Learn More
Med Vet International Yorker Dropper Bottle, 60mL, 12/Box
Catalog No. 502522482
Encompass_Preferred
Removeable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene. 12 per box.
Catalog No. 50-252-2482 Supplier Med Vet International Supplier No. 975060
Removeable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene. 12 per box.