Learn More
Beckman Coulter BOTTLE & CAP AY PC 5/8 X 3 (BOX 6)
Catalog No. NC9491225
Encompass_Preferred
Bottle Assembly, Polycarbonate, 10.4mL, 16x76mm, Quantity: 6/Box
Catalog No. NC9491225 Supplier Beckman Coulter Supplier No. 355603
Bottle Assembly, Polycarbonate, 10.4mL, 16x76mm, Quantity: 6/Box