Learn More
Description
BD BBL™ Tryptic Soy Broth is a ready-for-use prepared bottled media.
- Used in qualitative procedures for detection, isolation, and cultivation of fastidious and nonfastidious microorganisms.
- Applications include testing pharmaceutical, food, environmental, or water testing samples.
Specifications
Specifications
| Broth Type | Tryptic Soy Broth |
| Country of Origin | United States |
| Detects | Microorganisms |
| Form | Liquid |
| Format | Bottle |
| Manufacturing Origin | United States |
| Media Type | Broth |
| Preparation Method | Ready-to-Use |
| Product Type | Broth |
| Fill Volume | 200 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.