Learn More
Research Products International Corp Bottles, HDPE, 32oz, 6/pk
SDP
Catalog No. 501978274
Encompass
Precisionware Narrow-Mouth Bottles are made from rigid high-density polyethylene. These laboratory/pharmaceutical grade bottles have matched polypropylene closures and leak proof design. They are ideal for packaging and storage of liquids and powders. Bottles feature recycling code molded into the base.
Catalog No. 50-197-8274 Supplier Research Products International Corp Supplier No. F106200008
Precisionware Narrow-Mouth Bottles are made from rigid high-density polyethylene. These laboratory/pharmaceutical grade bottles have matched polypropylene closures and leak proof design. They are ideal for packaging and storage of liquids and powders. Bottles feature recycling code molded into the base.