Learn More
Beckman Coulter BOTTLES PP 4 X 6-1/2 (BOX-6)
Catalog No. NC9671807
Encompass_Preferred
Polypropylene Bottles with Screw Caps, Quantity 6, Volume Per Tube 1000mL, Size 97 x 167mm
This item is not returnable. View return policy
Catalog No. NC9671807 Supplier Beckman Coulter Supplier No. 355676
Polypropylene Bottles with Screw Caps, Quantity 6, Volume Per Tube 1000mL, Size 97 x 167mm