Learn More
Chemglass Life Sciences Bottletop Dispenser, Analog-Adjustable, 2.5-25mL, Dispensette S, with Recirculation Valve
SDP
Catalog No. 501531849
Encompass_Preferred
Bottletop Dispenser, Analog-Adjustable, 2.5-25mL, Dispensette S, with Recirculation Valve. Dispensette S bottletop dispensers are suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents. Extremely easy filling and dispensing. Simple priming by optimized flow channels. Rapid and safe volume adjustment with internal scalloped track. Dispensette S Analog-adjustable: Rapid and safe volume setting withinternal scalloped track. Simple calibration adjustment with a supplied tool.
Catalog No. 50-153-1849 Supplier Chemglass Life Sciences Supplier No. CG900125A
Bottletop Dispenser, Analog-Adjustable, 2.5-25mL, Dispensette S, with Recirculation Valve. Dispensette S bottletop dispensers are suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents. Extremely easy filling and dispensing. Simple priming by optimized flow channels. Rapid and safe volume adjustment with internal scalloped track. Dispensette S Analog-adjustable: Rapid and safe volume setting withinternal scalloped track. Simple calibration adjustment with a supplied tool.