Learn More
Cole-Parmer Cole-Parmer Bottletop Dispenser; 1 to 10 mL
Supplier:  Cole-Parmer 2520806
Encompass_Preferred
Unique PTFE piston with intergral o-ring provides smooth motion and high accuracy. Features a built-in 30-mm threaded screw top and include adapters for 28-, 32-, 36-, 40- and 45-mm bottles. Dispenser is easy to calibrate and disassemble for cleaning.
Catalog No. NC0139130