Learn More
Synthware™ Bottom Adapter for Vacuum Trap Body
GREENER_CHOICESDP
Catalog No. 31501801 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
31-501-801 1 Ea.
1 options
Catalog No. 31-501-801 Supplier Synthware™ Supplier No. V175039
Add to Cart
Add to Cart
Spherical joints add flexibility to the Vacuum Trap.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Product Type Bottom Adapter
Quantity 1 Ea.
Standard Taper Joint No. 35/20
Width (Metric) 12 mm