Learn More
Innovive Inc Mouse cage bottom for use with Innovive IVC or static mouse lids, prefilled with Alpha-dri bedding with paper enrichment added
Catalog No. NC2021501
Encompass_Preferred
Bottom, Alpha Dri w/ Innorichment (3ply)
Catalog No. NC2021501 Supplier Innovive Inc Supplier No. MBTMADE
Bottom, Alpha Dri w/ Innorichment (3ply)