Learn More
Ace Glass, Inc. Bottom Stopcock Adapter w/Kalrez o-ring 0-5mm s/c #15 thd for back sealing with 5857-101 adapter & #7 outlet
SDP
Catalog No. 5000810363
Encompass_Preferred
Bottom Stopcock Adapter w/Kalrez o-ring 0-5mm s/c #15 thd for back sealing with 5857-101 adapter & #7 outlet
Catalog No. 50-008-10363 Supplier Ace Glass, Inc. Supplier No. 583981
Bottom Stopcock Adapter w/Kalrez o-ring 0-5mm s/c #15 thd for back sealing with 5857-101 adapter & #7 outlet