Learn More
BUCHI Bottom Valve, Dn 25
SDP
Catalog No. 05405220 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
R-220 Pro, R-220 Essential, R-220 EX
Catalog No. For Use With (Equipment)
05-405-220 R-220 Pro, R-220 Essential, R-220 EX
1 options
Catalog No. 05-405-220 Supplier BUCHI Supplier No. 041061
Add to Cart
Add to Cart
Constructed of borosilicate glass 3.3, for use with draining receiving flasks.
    For use with draining receiving flasks
  • Compatible with R-220 Pro, R-220 Essential, and R-220 EX
Specifications
Description Dn 25
Material Borosilicate Glass (3.3)
Type Drain Valve
For Use With (Application) Draining Receiving Flask
For Use With (Equipment) R-220 Pro, R-220 Essential, R-220 EX