Learn More
Med Vet International Bouffant Cap, Spunbond, 21", Green, 500/Case, Cri1000
Catalog No. 502829633
Encompass_Preferred
Boundary Bouffant Caps: All caps comply with NFPA standards and are latex-free. Double-stitched elastic band keeps caps flat and comfortable on the forehead. Economy Cap. Lint Free. Made of Spunbond Polypropylene. Green, 21".
Catalog No. 50-282-9633 Supplier Med Vet International Supplier No. CRI1000
Boundary Bouffant Caps: All caps comply with NFPA standards and are latex-free. Double-stitched elastic band keeps caps flat and comfortable on the forehead. Economy Cap. Lint Free. Made of Spunbond Polypropylene. Green, 21".