Learn More
Fisherbrand™ Bouffant Cap and Beard Cover Hood
Catalog No. 128970041 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White
Catalog No. Color
12-897-0041 White
1 options
This item is not returnable. View return policy
Catalog No. 12-897-0041
This item is not returnable. View return policy
This is manufactured from a specially formulated spunbond polypropylene (SBPP) fabric, and can be used in cleanroom environments.
  • Economical, breatheable spunbound polypropylene fabric
  • One size fits most
  • Elastic around the facial opening and neck ensures a secure fit
Specifications
Product Type Combination Bouffant and Beard Cover
Size One Size
Material Spunbond Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Style Combination Bouffant and Beard Cover