Learn More
Grainger Action Chemical Bouffant Cap, Polypropylene, 24 in., Universal, 1000/Pk.
Catalog No. 502770907
Encompass_Preferred
These bouffant caps provide reliable head coverage for laboratory and cleanroom environments. Constructed from polypropylene, they offer a comfortable fit while maintaining a lint-free surface to prevent contamination.
  • Material: Polypropylene
  • Color: Blue
  • Diameter: 24 in.
  • Universal size
  • Latex-free
  • Lint-free construction
  • Packaging: 1,000 per box
  • Complies with FDA regulations
Catalog No. 50-277-0907 Supplier Grainger Supplier No. 13V912
These bouffant caps provide reliable head coverage for laboratory and cleanroom environments. Constructed from polypropylene, they offer a comfortable fit while maintaining a lint-free surface to prevent contamination.
  • Material: Polypropylene
  • Color: Blue
  • Diameter: 24 in.
  • Universal size
  • Latex-free
  • Lint-free construction
  • Packaging: 1,000 per box
  • Complies with FDA regulations