Learn More
Grainger ACTION CHEMICAL Bouffant Cap, Polypropylene, Green, 24 in. Diameter, 1,000/Pk.
Catalog No. 502770211
Encompass_Preferred
These green bouffant caps are designed for reliable hair coverage in controlled environments. Constructed from polypropylene, these caps provide a lightweight and comfortable fit for extended use.
  • Diameter: 24 in.
  • Material: Polypropylene
  • Latex-free construction
  • Lint-free properties
  • Size: L, universal
  • Color: Green
  • Package quantity: 1,000
Catalog No. 50-277-0211 Supplier Grainger Supplier No. 13V915
These green bouffant caps are designed for reliable hair coverage in controlled environments. Constructed from polypropylene, these caps provide a lightweight and comfortable fit for extended use.
  • Diameter: 24 in.
  • Material: Polypropylene
  • Latex-free construction
  • Lint-free properties
  • Size: L, universal
  • Color: Green
  • Package quantity: 1,000