Learn More
Grainger Action Chemical Bouffant Cap, Polypropylene, Red, 21 in. Diameter, 1,000/Pk.
Catalog No. 502773174
Encompass_Preferred
These bouffant caps provide lightweight and breathable head protection for cleanroom or clinical environments. Constructed from polypropylene, the caps offer a comfortable fit while maintaining barrier protection.
  • Constructed from durable polypropylene
  • Lint-free material helps maintain a clean environment
  • Latex-free construction prevents allergic reactions
  • Designed with a 21 in. diameter for a secure fit
  • Supplied in a box of 1,000 caps
Catalog No. 50-277-3174 Supplier Grainger Supplier No. 13V916
These bouffant caps provide lightweight and breathable head protection for cleanroom or clinical environments. Constructed from polypropylene, the caps offer a comfortable fit while maintaining barrier protection.
  • Constructed from durable polypropylene
  • Lint-free material helps maintain a clean environment
  • Latex-free construction prevents allergic reactions
  • Designed with a 21 in. diameter for a secure fit
  • Supplied in a box of 1,000 caps