Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Flat Bouffant Caps

Catalog No. 19472790 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Color:
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-472-790 White
19-472-789 White
19-472-791 -
19-472-792 White
19-472-793 -
19-472-794 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-472-790 Supplier International Enviroguard™ Supplier No. S110NW19
Add to Cart
Add to Cart

Flat bouffant caps provide protection against dirt, grime, and particulates in non-hazardous environments. Available in blue or white in a variety of sizes.

  • Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
  • Low-linting, lightweight cover restrains hair and offers great ventilation
  • Designed to protect against dirt, grime, and particulates in non-hazardous environments
  • Flat style
  • Latex-free elastic
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more

Specifications

Product Type Bouffant
Material Polypropylene
Color White
Disposable Single-use
Model S110NW
Diameter (English) 19 in.
For Use With (Application) Food Processing, Construction, Pharmaceuticals, Environmental Cleanup
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.