Learn More
VGuard™ Bouffant Caps
Description
- Strong, elastic headband
- Superior barrier properties
- Available in pancake or pleated styles
Specifications
Specifications
| Material | Spunbond Polypropylene |
| Color | Green |
| Disposable | Single-use |
| Model | B11A62 |
| For Use With (Application) | Food Service, Food Processing, Industrial, Janitorial, Manufacturing |
| Style | Pancake |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.